missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB513-SP
This item is not returnable.
View return policy
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| MMP-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4314 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 50647 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P08254 | |
| MMP3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-3 Tyr18-Cys477 Accession # P08254 | |
| 25 μg | |
| Primary | |
| Detects both the pro and active forms of human MMP-3 in direct ELISAs and Western blots. In direct ELISAs, 50-100% cross-reactivity with recombinant mouse MMP-3 is observed, 10% cross-reactivity with recombinant human (rh) MMP-10 is observed and no cross-reactivity with rhMMP-1, -2, -7, -8, -9, -12 or -13 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction