missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-25/MT6-MMP Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1142
Additional Details : Weight : 0.00970kg
Description
MMP-25/MT6-MMP Monoclonal specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
MMP-25/MT6-MMP | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9NPA2 | |
MMP25 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-25/MT6-MMP Pro314-Cys508 Accession # Q9NPA2 | |
500 μg | |
Primary | |
Detects human MMP-25/MT6-MMP in Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
141808 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP25, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64386 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only