missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9024-SP
This item is not returnable.
View return policy
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human samples. It is validated for ELISA.
Specifications
| MMP-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Chinese hamster ovary cell line, CHO-derived recombinant human MMP-2 Ile34-Cys660 Accession # P08253 | |
| 25 μg | |
| 4313 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 666047 | |
| ELISA | |
| P08253 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human MMP-2 in direct ELISAs. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction