missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-19 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF6790-SP
This item is not returnable.
View return policy
Description
MMP-19 Polyclonal specifically detects MMP-19 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MMP-19 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24.-, matrix metallopeptidase 19, matrix metalloproteinase 18, matrix metalloproteinase 19, Matrix metalloproteinase RASI, Matrix metalloproteinase-18, matrix metalloproteinase-19, MMP-18, MMP18MMP-19, MMP19, RASI, RASI-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4327 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q99542 | |
| MMP19 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-19 Leu229-Tyr508 Accession # Q99542 | |
| 25 μg | |
| Primary | |
| Detects human MMP-19 in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction