missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-15/MT2-MMP Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB9161
This item is not returnable.
View return policy
Description
MMP-15/MT2-MMP Monoclonal specifically detects MMP-15/MT2-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| MMP-15/MT2-MMP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MMP15 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-15 aa 132-625 | |
| 500 μg | |
| Primary | |
| Detects human MMP-15/MT2-MMP in direct ELISAs and Western blots. Does not detect the catalytic domain of E. coli-expressed recombinant human MMP-15 (aa 132-304). | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 130522 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 15 (membrane-inserted), matrix metalloproteinase 15 (membrane-inserted), Membrane-type matrix metalloproteinase 2, Membrane-type-2 matrix metalloproteinase, MMP15, MMP-15, MT2-MMP, MT2-MMPMT2MMP, MTMMP2MT-MMP 2, SMCP-2matrix metalloproteinase-15 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4324 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction