missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-14/MT1-MMP Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB918
This item is not returnable.
View return policy
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| MMP-14/MT1-MMP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP14, MMP-14, MMP-X1, MT1MMP, MT1-MMP, MT1-MMPMTMMP1, MT-MMP 1, MT-MMP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4323 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 5H2 | |
| Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Immunoprecipitation 25 μg/mL | |
| P50281 | |
| MMP14 | |
| Human MMP-14 synthetic peptide AYIREGHEKQA Accession # P50281 | |
| 500 μg | |
| Primary | |
| Detects recombinant human (rh) MMP-14 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) MMP-1, -2, -7, -8, -9, -10, -12, -13 or the catalytic domains of rhMMP-15 and -16. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction