missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MMP-14 ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.
Specifications
Specifications
| Accession Number | P50281 |
| Assay Range | 0.63 to 40 ng/mL |
| Assay Sensitivity | 0.38 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 4323 |
| Gene Symbol | MMP14 |
| Interassay CV | <10% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction