missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-11 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC3657R-100UG
This item is not returnable.
View return policy
Description
MMP-11 Monoclonal specifically detects MMP-11 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MMP-11 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P24347 | |
| MMP11 | |
| E. coli-derived recombinant human MMP-11 Met1-Leu488 Accession # P24347 | |
| RUO | |
| 4320 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 135421 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MMP-11 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction