missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-10 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9102
This item is not returnable.
View return policy
Description
MMP-10 Monoclonal specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| MMP-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4319 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF | |
| 110316 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P09238 | |
| MMP10 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-10 Phe99-Cys476 Accession # P09238 | |
| 500 μg | |
| Primary | |
| Detects the pro and active forms of human MMP-10 in direct ELISAs Western blots. In direct ELISAs and Western blots, approximately 10-50% cross-reactivity with recombinant human (rh) MMP-3 and no cross-reactivity with rhMMP-1, -2, -7, -8, -9, -12, or -13 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction