missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human MMP-1 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 250 μg
2 x 500 μg
250 μg
50 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
Description
420-01-1MG will be provided as 4 x 250 μg (420-01-250UG) 420-01-500UG will be provided as 2 x 250 μg (420-01-250UG) Recombinant Human MMP-1 is a 42.7 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain, which is involved in substrate specificity, and in binding TIMP-1. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | P03956 |
| For Use With (Application) | Control, Enzyme Activity, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 4312 |
| Molecular Weight (g/mol) | 42.7 kDa |
| Name | Human MMP-1 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction