missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF901-SP
This item is not returnable.
View return policy
Description
MMP-1 Polyclonal specifically detects MMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated, ELISA Capture (Matched Antibody Pair).
Specifications
| MMP-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4312 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, KnockDown, ELISA | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| P03956 | |
| MMP1 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-1 Phe20-Asn469 Accession # P03956 | |
| 25 μg | |
| Primary | |
| Detects human MMP-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.5% cross-reactivity with recombinant human (rh) MMP-2, rhMMP-3, rhMMP-7, rhMMP-8, rhMMP-9, rhMMP-10, rhMMP-12, rhMMP-13, and rhMMP-16 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction