missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MMACHC (aa 197-279) Control Fragment Recombinant Protein

Product Code. 30195194
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195194

Brand: Invitrogen™ RP94201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55596 (PA5-55596. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y4U1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25974
Name Human MMACHC (aa 197-279) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810037K07Rik; cblC; Cyanocobalamin reductase (cyanide-eliminating); methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria; methylmalonic aciduria and homocystinuria type C protein; Methylmalonic aciduria and homocystinuria type C protein homolog; methylmalonic aciduria cblC type, with homocystinuria; Mmachc; RGD1310806; RP11-291L19.3
Common Name MMACHC
Gene Symbol MMACHC
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NFHWRDWTYRDAVTPQERYSEEQKAYFSTPPAQRLALLGLAQPSEKPSSPSPDLPFTTPAPKKPGNPSRARSWLSPRVSPPAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.