missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MLLT1 (aa 142-260) Control Fragment Recombinant Protein

Product Code. 30209502
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209502

Brand: Invitrogen™ RP102514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83183 (PA5-83183. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Chromosome band 11q23 is the site of translocations in myeloid and lymphoid acute leukemias, pediatric leukemias, and treatment-induced secondary acute myelogenous leukemia. The translocation breakpoints cluster in a restricted region of the HRX gene resulting in chimeric genes that encode an N-terminal portion of Hrx fused to various partner proteins. Myeloid/lymphoid or mixed-lineage leukemia translocated to 1 (MLLT1) is a nuclear protein with transcriptional transactivation properties that is fused to Hrx in t(11;19) leukemias. The minimal MLLT1 sequence required for transcription activation was narrowed to the C-terminal 90 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03111
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4298
Name Human MLLT1 (aa 142-260) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA407901; BAM11; CTC-503J8.6; ENL; ENL/MLL fusion; leukemia-associated protein MLLT1; LTG19; MLL/ENL fusion protein; MLLT1; MLLT1 super elongation complex subunit; MLLT1, super elongation complex subunit; MLLT1/MLL fusion; myeloid/lymphoid or mixed lineage leukemia translocation to 1 homolog; myeloid/lymphoid or mixed lineage-leukemia translocation to 1 homolog; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 1; protein ENL; trithorax; YEATS domain-containing protein 1; YEATS1
Common Name MLLT1
Gene Symbol MLLT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MVMPEGADTVSRPSPDYPMLPTIPLSAFSDPKKTKPSHGSKDANKESSKTSKPHKVTKEHRERPRKDSESKSSSKELEREQAKSSKDTSRKLGEGRLPKEEKAPPPKAAFKEPKMALKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.