missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MLK3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4559
This item is not returnable.
View return policy
Description
MLK3 Polyclonal specifically detects MLK3 in Human samples. It is validated for Western Blot.
Specifications
| MLK3 | |
| Polyclonal | |
| Unconjugated | |
| Q16584 | |
| MAP3K11 | |
| E. coli-derived recombinant human MLK3 Ala85-Gly207 Accession # Q16584 | |
| 100 μg | |
| Primary | |
| Detects endogenous human MLK3 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| EC 2.7.11, EC 2.7.11.25, MAP3K11, MEKK11, mitogen-activated protein kinase kinase kinase 11, Mixed lineage kinase 3, MLK3MLK-3, PTK1, PTK1protein-tyrosine kinase PTK1, RHOE, SH3 domain-containing proline-rich kinase, SPRK, SPRKMGC17114, Src-homology 3 domain-containing proline-rich kinase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4296 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction