missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MKP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB35761-SP
This item is not returnable.
View return policy
Description
MKP-3/DUSP6 Monoclonal specifically detects MKP-3/DUSP6 in Human samples. It is validated for Immunocytochemistry.
Specifications
| MKP-3/DUSP6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, DUSP6, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1, PYST1dual specificity protein phosphatase 6 | |
| E. coli-derived recombinant human MKP-3, Met1-Thr381, Accession No. Q16828 | |
| Primary | |
| Detects human MKP-3 in direct ELISAs. | |
| Human | |
| IgG2a |
| Immunocytochemistry | |
| 363209 | |
| Immunocytochemistry 8-25 μ/mL | |
| Q16828 | |
| Mouse | |
| 25 μg | |
| 1848 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction