missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MKP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7168-SP
This item is not returnable.
View return policy
Description
MKP-1/DUSP1 Monoclonal specifically detects MKP-1/DUSP1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| MKP-1/DUSP1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P28562 | |
| DUSP1 | |
| E. coli-derived recombinant human MKP-1 Met1-Cys367 Accession # P28562 | |
| 25 μg | |
| Primary | |
| Detects human MKP-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) MKP-2, rhMKP-3, or rhMKP-5 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 408612 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CL100, CL100MAP kinase phosphatase 1, dual specificity phosphatase 1, Dual specificity protein phosphatase hVH1, DUSP1, EC 3.1.3.16, EC 3.1.3.48, HVH1, Mitogen-activated protein kinase phosphatase 1, MKP1, MKP-1dual specificity protein phosphatase 1, Protein-tyrosine phosphatase CL100, PTPN10, PTPN10serine/threonine specific protein phosphatase, VH1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1843 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction