missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MIS/AMH ELISA Kit

Product Code. 17747703
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17747703 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17747703 Supplier Invitrogen™ Supplier No. EH326RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P03971
Assay Range 0.18 to 40 ng/mL
Assay Sensitivity 0.18 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 268
Gene Symbol AMH
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target AMH
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.