Learn More
Invitrogen™ Human MIP-1b (CCL4) Instant ELISA™ Kit
Description
The Human Macrophage Inflammatory Protein-1beta (CCL4)(Hu MIP-1β) ELISA quantitates Hu MIP-1β in human serum, plasma, cell culture supernatants or other body fluids. The assay will exclusively recognize both natural and recombinant Hu MIP-1β. Principle of the method The Human MIP-1β solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P13236 |
| Assay Range | 7.8-500 pg/mL |
| Assay Sensitivity | 3.0 pg/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with Polyclonal Antibody to MIP-1beta, Biotin-Conjugate (anti-MIP-1beta polyclonal antibody), Streptavidin-HRP and Sample diluent, lyophilized Aluminum pouch(es) with a human MIP-1beta Standard curve (coloured) Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Sample Diluent (Use when an external predilution of the samples is needed) Stop Solution (1M Phosphoric acid) Adhesive Plate Covers |
| Conjugate | HRP |
| Immunoassay Kit Format | Coated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.