missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MIP-1 delta (aa 87-113) Control Fragment Recombinant Protein

Product Code. 30201494
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201494

Brand: Invitrogen™ RP106873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66186 (PA5-66186. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16663
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6359
Name Human MIP-1 delta (aa 87-113) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2 Lkn-1; HMRP-2 B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1 D; MIP5; MIP-5; MIP-5 MIP-1 d; Mrp-2 b; NCC3; NCC-3; NCC-3 MIP-1 D; NCC3MRP-2 B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15
Common Name MIP-1 delta
Gene Symbol CCL15
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KKGRQVCAKPSGPGVQDCMKKLKPYSI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.