missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mindin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £337.00
Specifications
| Antigen | Mindin/Spondin-2 |
|---|---|
| Clone | 331620 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715749
![]() |
R&D Systems
MAB2609-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141501
|
R&D Systems
MAB2609 |
100 μg |
£337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mindin/Spondin-2 Monoclonal specifically detects Mindin/Spondin-2 in Human samples. It is validated for Western Blot.Specifications
| Mindin/Spondin-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAH36341 | |
| 10417 | |
| Mouse myeloma cell line NS0-derived recombinant human Mindin Glu32-Val331 Accession # AAH36341 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Mindin in direct ELISAs and Western blots. |
| 331620 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Differentially expressed in cancerous and non-cancerous lung cells 1, DIL1, DIL-1, DIL1FLJ34460, DKFZp686G21139, FLJ16313, FLJ22401, MINDIN, M-SPONDIN, RG-1, SPON2, Spondin 2, spondin 2, extracellular matrix protein, spondin-2 | |
| SPON2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title