missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mind Bomb 2/MIB2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB72891-SP
This item is not returnable.
View return policy
Description
Mind Bomb 2/MIB2 Monoclonal specifically detects Mind Bomb 2/MIB2 in Human samples. It is validated for Western Blot.
Specifications
| Mind Bomb 2/MIB2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q96AX9 | |
| MIB2 | |
| E. coli-derived recombinant human Mind Bomb 2/MIB2 Ala418-Lys520 Accession # Q96AX9 | |
| 25 μg | |
| Primary | |
| Detects human Mind Bomb 2/MIB2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 739505 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, MIB2, Mind bomb homolog 2, Mind Bomb-2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1, ZZZ5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 142678 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction