missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MINA53 (aa 1-148) Control Fragment Recombinant Protein

Product Code. 30212541
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212541

Brand: Invitrogen™ RP89697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MINA is nuclear localized, myc-inducible protein that is thought to play a role in mammalian cell proliferation. Treatment of cancer cells lines such as the colon cancer cell line SW680 with siRNA against MINA inhibits cell growth, demonstrating that MINA may be a potential therapeutic target. MINA regulates several genes related to cell adhesion and metabolism that have also been shown to be regulated by c-Myc, but also regulates other genes whose expression are not modulated by c-Myc such as EGFR, IL-6 and HGF. MINA has also been found to act as a repressor to IL-4 expression in T cells, indicating that it may also play a role in T cell differentiation and genetic variation in T helper type 2 bias.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IUF8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84864
Name Human MINA53 (aa 1-148) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810047J07Rik; 2410057H13Rik; 3830408E23Rik; 60 S ribosomal protein L27a histidine hydroxylase; AI449204; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; DKFZp762O1912; FLJ14393; histone lysine demethylase MINA; MDIG; mina; mina {ECO:0000312; Mina53; Mina-53; mineral dust induced gene protein; mineral dust-induced gene protein; MYC induced nuclear antigen; myc-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; NO52; Nucleolar protein 52; Protein mina53; RGD:708521}; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; RIO x 2; ROX
Common Name MINA53
Gene Symbol RIOX2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPKKAKPTGSGKEEGPAPCKQMKLEAAGGPSALNFDSPSSLFESLISPIKTETFFKEFWEQKPLLIQRDDPALATYYGSLFKLTDLKSLCSRGMYYGRDVNVCRCVNGKKKVLNKDGKAHFLQLRKDFDQKRATIQFHQPQRFKDELW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.