missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MIF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 16 publications
Brand: R&D Systems MAB289-100
This item is not returnable.
View return policy
Description
MIF Monoclonal specifically detects MIF in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| MIF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4282 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 12302 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P14174 | |
| MIF | |
| E. coli-derived recombinant human MIF Pro2-Ala115 Accession # P14174 | |
| 100 μg | |
| Primary | |
| Detects human MIF in Western blots. In Western blots, this antibody shows approximately 25% cross-reactivity with recombinant mouse MIF. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction