missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Midkine Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC2581V-100UG
This item is not returnable.
View return policy
Description
Midkine Monoclonal specifically detects Midkine in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Midkine | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| E. coli-derived recombinant human Midkine., Lys23-Asp143, Accession No. P21741 | |
| 100 μg | |
| 4192.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2284A | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P21741 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Midkine in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction