missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MID1IP1 (aa 62-136) Control Fragment Recombinant Protein

Product Code. 30198393
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30198393 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30198393 Supplier Invitrogen™ Supplier No. RP95456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58552 (PA5-58552. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPA3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58526
Name Human MID1IP1 (aa 62-136) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3110038L01Rik; FLJ10386; G12-like; gastrulation specific G12 homolog; gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1; MID1 interacting protein 1 (gastrulation specific G12 homolog); Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)); MID1 interacting protein 1 (gastrulation specific G12-like); mid1-interacting G12-like protein; Mid1-interacting protein 1; MID1IP1; mid1ip1 {ECO:0000250; Mig12; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; slap; spot 14-like androgen-inducible protein; spot 14-R; spot 14-related protein; STRAIT11499; THRSPL; UniProtKB:Q9CQ20}
Common Name MID1IP1
Gene Symbol MID1IP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CLEERTPPVPDSGSANGSFFAPSRDMYSHYVLLKSIRNDIEWGVLHQPPPPAGSEEGSAWKSKDILVDLGHLEGA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.