missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Microsomes, 50 Donors

Product Code. 17155989
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.50mL
This item is not returnable. View return policy

Product Code. 17155989

Brand: Gibco™ HMMCPL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human Microsomes, 50 Donors

Human liver microsomes pooled from 50 different individual donors. All donors are equally represented for a truer population sample. Fully characterized for major cytochrome P450 activities and select Phase II enzymes.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
TRUSTED_SUSTAINABILITY

Specifications

Donor Source Pooled
Gender Unisex
Physical Form Frozen
Tissue Liver
Quantity 0.5 mL
Purity Research Grade
Host Species Human
Concentration 20 mg/mL
Product Type Human Microsome
Content And Storage Store in ultracold freezer (-60 to -85°C) and protect from light.
Life Stage Adult
Format Tube
Sub Cellular Localization Microsomes
Shipping Condition Dry Ice
Product Line Gibco
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.