missing translation for 'onlineSavingsMsg'
Learn More

GIBCO™ Human Microsomes, 20 Donors, Female Donors

Product Code. 10690459
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10690459 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10690459 Supplier GIBCO™ Supplier No. HMMCPF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human liver microsomes pooled from 20 different individual donors. All donors are equally represented for a truer population sample. These microsomes have been fully characterized for major cytochrome P450 activities and select Phase II enzymes.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Target-Based ADME/Tox Assays, Biochemical ADME Assays, Enzymes and Proteins for ADME, Hepatocytes and Hepatic Models, Industrial and Applied Science, Pharma and Biopharma
Tissue Liver
Quantity 0.5 mL
Cellular Fraction Microsomes
Host Species Human
Concentration 20 mg/mL
Content And Storage Store between -60°C and -85°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.