missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1599-SP
This item is not returnable.
View return policy
Description
MICB Monoclonal specifically detects MICB in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated, ELISA Capture (Matched Antibody Pair).
Specifications
| MICB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CAI18747 | |
| MICB | |
| Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 | |
| 25 μg | |
| Primary | |
| Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF, Immunoassay, ELISA | |
| 236511 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4277 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction