missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICB Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£210.00 - £469.00
Specifications
| Antigen | MICB |
|---|---|
| Clone | 236511 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743759
![]() |
R&D Systems
FAB1599G-025 |
25 Tests |
£210.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18154611
|
R&D Systems
FAB1599G-100 |
100 Tests |
£469.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICB Monoclonal specifically detects MICB in Human samples. It is validated for Flow Cytometry.Specifications
| MICB | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
| MICB | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA. |
| 236511 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| CAI18747 | |
| 4277 | |
| Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title