missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MICAL3 (aa 1297-1440) Control Fragment Recombinant Protein

Product Code. 30210467
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210467

Brand: Invitrogen™ RP91479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51431 (PA5-51431. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Monooxygenase that promotes depolymerization of F-actin by mediating oxidation of specific methionine residues on actin to form methionine-sulfoxide, resulting in actin filament disassembly and preventing repolymerization. In the absence of actin, it also functions as a NADPH oxidase producing H(2)O(2). Seems to act as Rab effector protein and plays a role in vesicle trafficking. Involved in exocytic vesicles tethering and fusion: the monooxygenase activity is required for this process and implicates RAB8A associated with exocytotic vesicles. Required for cytokinesis. Contributes to stabilization and/or maturation of the intercellular bridge independently of its monooxygenase activity. Promotes recruitment of Rab8 and ERC1 to the intercellular bridge, and together these proteins are proposed to function in timely abscission. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7RTP6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57553
Name Human MICAL3 (aa 1297-1440) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias [F-actin]-monooxygenase MICAL3; flavoprotein oxidoreductase MICAL3; KIAA0819; KIAA1364; MICAL3; MICAL-3; microtubule associated monooxygenase, calponin and LIM domain containing 3; molecule interacting with CasL protein 3; protein MICAL-3; protein-methionine sulfoxide oxidase MICAL3
Common Name MICAL3
Gene Symbol MICAL3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PVQSQSDTKDRLGSPLAVDEALRRSDLVEEFWMKSAEIRRSLGLTPVDRSKGPEPSFPTPAFRPVSLKSYSVEKSPQDEGLHLLKPLSIPKRLGLPKPEGEPLSLPTPRSPSDRELRSAQEERRELSSSSGLGLHGSSSNMKTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.