missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MICAL2 (aa 592-718) Control Fragment Recombinant Protein

Product Code. 30202392
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202392

Brand: Invitrogen™ RP88764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56474 (PA5-56474. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MICALs (Molecules Interacting with CasL) are atypical multidomain flavoenzymes with diverse cellular functions. There are three known isoforms, MICAL1, MICAL2 and MICAL3, as well as the MICAL-like proteins MICAL-L1 and MICAL-L2. MICAL2 has three conserved domains: an N-terminal flavin adenine dinucleotide (FAD) binding domain, a calponin homology (CH) domain and a Lin11, Isl-1 and Mec-3 (LIM) domain. It has been demonstrated that MICAL2 could regulate actin stress fibers and is required for normal actin organization. In addition, MICAL2-PV, a novel splicing variants of MICAL2, has been reported to be involved in cancer progression of prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94851
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9645
Name Human MICAL2 (aa 592-718) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias [F-actin]-monooxygenase MICAL2; 5330438E18Rik; 9530064J02; flavoprotein oxidoreductase MICAL2; Kiaa0750; Mical2; MICAL-2; MICAL2PV1; MICAL2PV2; microtubule associated monooxygenase, calponin and LIM domain containing 2; microtubule associated monoxygenase, calponin and LIM domain containing 2; mKIAA0750; mMical2; molecule interacting with CasL protein 2; protein MICAL-2; protein-methionine sulfoxide oxidase MICAL2; RGD1311773
Common Name MICAL2
Gene Symbol Mical2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PVTTGKEMASAQEPDKLSMVMYLSKFYELFRGTPLRPVDSWRKNYGENADLSLAKSSISNNYLNLTFPRKRTPRVDGQTGENDMNKRRRKGFTNLDEPSNFSSRSLGSNQECGSSKEGGNQNKVKSM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.