missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICA/B PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
£198.00 - £480.00
Specifications
| Antigen | MICA/B |
|---|---|
| Clone | 159207 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756867
![]() |
R&D Systems
FAB13001P-025 |
25 Tests |
£198.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729896
![]() |
R&D Systems
FAB13001P-100 |
100 Tests |
£480.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICA/B Monoclonal specifically detects MICA/B in Human samples. It is validated for Flow Cytometry.Specifications
| MICA/B | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
| MICA | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human MICA/B in direct ELISAs. |
| 159207 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| Q29983 | |
| 100507436 | |
| BaF3 mouse pro-B cell line transfected with human MICA Accession # Q29983 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title