missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICA/B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
£178.00 - £1755.00
Specifications
| Antigen | MICA/B |
|---|---|
| Clone | 159207 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753348
![]() |
R&D Systems
MAB13001-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772618
![]() |
R&D Systems
MAB13001-100 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759437
![]() |
R&D Systems
MAB13001-500 |
500 μg |
£1755.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICA/B Monoclonal specifically detects MICA/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| MICA/B | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
| MICA | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 159207 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q29983 | |
| 100507436 | |
| BaF3 mouse pro-B cell line transfected with human MICA Accession # Q29983 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MICA/B in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title