missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MIA-2 Recombinant Protein, PeproTech®

Product Code. 17820373
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17820373

Brand: Gibco™ 130021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MIA-2 is a 101 amino acid, 11.5 kDa protein comprising the MIA homologous N-terminal region of the full length MIA-2 protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PC5
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4253
Molecular Weight (g/mol) 11.5 kDa
Name Human MIA-2
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cTAGE family member 5; CTAGE family member 5 ER export factor; CTAGE family member 5, ER export factor; CTAGE family, member 5; CTAGE5; cutaneous T-cell lymphoma-associated antigen 5; LOW QUALITY PROTEIN: melanoma inhibitory activity protein 2; MEA11; MEA6; melanoma inhibitory activity 2; melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); meningioma-expressed antigen 6/11; MGEA; MGEA11; MGEA6; MIA SH3 domain ER export factor 2; MIA2; protein cTAGE-5
Common Name CTAGE5
Gene Symbol Mia2
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLESTKLLAD LKKCGDLECE ALINRVSAMR DYRGPDCRYL NFTKGEEISV YVKLAGERED LWAGSKGKEF GYFPRDAVQI EEVFISEEIQ MSTKESDFLC L
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.