missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £386.00
Specifications
| Antigen | mGluR3 |
|---|---|
| Clone | 440511 |
| Dilution | Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724177
![]() |
R&D Systems
MAB46682-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772037
![]() |
R&D Systems
MAB46682-100 |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR3 Monoclonal antibody specifically detects mGluR3 in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
| mGluR3 | |
| Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| GLUR3, glutamate receptor, metabotropic 3, GPRC1C, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
| Hek293 cells transfected with human mGLUR3, Accession # Q14832 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 440511 | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q14832 | |
| 2913 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title