missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £406.00
Specifications
| Antigen | mGluR3 |
|---|---|
| Clone | 530510 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753997
![]() |
R&D Systems
MAB46681-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18793747
![]() |
R&D Systems
MAB46681-100 |
100 μg |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR3 Monoclonal specifically detects mGluR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| mGluR3 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| GLUR3, glutamate receptor, metabotropic 3, GPRC1C, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human mGluR3, Asp25-Ser507, Accession # Q14832 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 530510 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q14832 | |
| 2913 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title