missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR1a Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF4836
Additional Details : Weight : 0.00970kg
Description
mGluR1 Polyclonal specifically detects mGluR1 in Human samples. It is validated for Western Blot.Specifications
mGluR1 | |
Polyclonal | |
Unconjugated | |
Q13255 | |
GRM1 | |
Chinese hamster ovary cell line CHO-derived recombinant human mGluR1 Ser33-Ser522 Accession # Q13255 | |
100 μg | |
Primary | |
Detects human mGluR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) mGluR4, rhmGluR8, rhmGluR2, rhmGluR3 and rhmGluR7 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
glutamate receptor, metabotropic 1, GPRC1A, GRM1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
Sheep | |
Affinity Purified | |
RUO | |
2911 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only