missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MGAT4B (aa 35-123) Control Fragment Recombinant Protein

Product Code. 30208402
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208402

Brand: Invitrogen™ RP101404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62561 (PA5-62561. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Has lower affinities for donors or acceptors than MGAT4A, suggesting that, under physiological conditions, it is not the main contributor in N- glycan biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UQ53
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11282
Name Human MGAT4B (aa 35-123) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2.4.1.145; alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase B; alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; aminyltransferase; glcNAc-T IVb; GNT-IV; GNT-IVB; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isoenzyme B; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B; MGAT4B; N-acetylglucosaminyltransferase IVb; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVb; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVb; UNQ906/PRO1927
Common Name MGAT4B
Gene Symbol MGAT4B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.