missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MFSD2B (aa 478-503) Control Fragment Recombinant Protein

Product Code. 30181964
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181964

Brand: Invitrogen™ RP99710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60628 (PA5-60628. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Multidrug transporters, such as MFSD2B, are membrane proteins that expel a wide spectrum of cytotoxic compounds from the cell and render cells resistant to multiple drugs. Major Facilitator Superfamily (MFS) members are capable of transporting various substrates such as sugars, polyols, drugs, neurotransmitters, amino acids, peptides, and inorganic anions, although most members are substrate-specific. MFSD2B is closely related to MFSD2A, which is expressed in many tissues and is highly induced in liver and brown adipose tissue (BAT) during fasting, playing a major role in the induction of MFSD2A expression during adaptive thermogenesis. Unlike MFSD2A, MFSD2B is expressed in few tissues and at relatively low levels, and is not induced during fasting, suggesting that MFSD2B may play other roles.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A6NFX1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 388931
Name Human MFSD2B (aa 478-503) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias hMfsd2b; major facilitator superfamily domain containing 2 B; major facilitator superfamily domain-containing protein 2 B; MFSD2B; RGD1561716
Common Name MFSD2B
Gene Symbol MFSD2B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VGSTPKTPSRDASSRLSLRRRTSYSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.