missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METTL24 (aa 287-365) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57953 (PA5-57953. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
METTL24 gene ontology annotations related to this gene include methyltransferase activity.
Specifications
Specifications
| Accession Number | Q5JXM2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 728464 |
| Name | Human METTL24 (aa 287-365) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 9030224M15Rik; C6orf186; C9H6orf186; dJ71D21.2; methyltransferase like 24; methyltransferase-like protein 24; Mettl24; UPF0624 protein C6orf186; UPF0624 protein C6orf186 homolog |
| Common Name | METTL24 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction