missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METTL11B (aa 216-277) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55753 (PA5-55753. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Alpha-N-methyltransferase that methylates the N-terminus of target proteins containing the N-terminal motif [Ala/Pro/Ser]-Pro-Lys when the initiator Met is cleaved. Specifically catalyzes monomethylation of exposed alpha-amino group of Ala or Ser residue in the [Ala/Ser]-Pro-Lys motif and Pro in the Pro-Pro-Lys motif. May activate NTMT1 by priming its substrates for trimethylation.
Specifications
Specifications
| Accession Number | Q5VVY1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 149281 |
| Name | Human METTL11B (aa 216-277) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | alpha N-terminal protein methyltransferase 1 B; C1orf184; Gm207; HOMT1B; methyltransferase like 11 B; methyltransferase-like protein 11 B; METTL11B; NRMT2; NTM1B; RGD1564106; X-Pro-Lys N-terminal protein methyltransferase 1 B |
| Common Name | METTL11B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction