missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Meteorin-like/METRNL Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £432.00
Specifications
| Antigen | Meteorin-like/METRNL |
|---|---|
| Clone | 2441D |
| Dilution | Immunohistochemistry 3-25 μ/mL, Immunocytochemistry 3-25 μ/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760857
![]() |
R&D Systems
MAB78672-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723627
![]() |
R&D Systems
MAB78672-100 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| Meteorin-like/METRNL | |
| Immunohistochemistry 3-25 μ/mL, Immunocytochemistry 3-25 μ/mL | |
| Monoclonal | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788 | |
| Chinese Hamster Ovary cell line, CHO-derived human Meteorin-like/METRNL, Gln46-Asp311, Accession No. Q641Q3 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2441D | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| Q641Q3 | |
| 284207 | |
| IgG | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Meteorin-like/METRNL in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title