missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MET (NM_000245.2, 956 a.a. - 1390 a.a.) F1200I mutant Partial Protein expressed in Sf9 cells

Product Code. 16008036
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16008036

Brand: Abnova™ P4825.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

The proto-oncogene MET product is the hepatocyte growth factor receptor and encodes tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. Various mutations in the MET gene are associated with papillary renal carcinoma. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KKRKQIKDLGSELVRYDARVHTPHLDRLVSARSVSPTTEMVSNESVDYRATFPEDQFPNSSQNGSCRQVQYPLTDMSPILTSGDSDISSPLLQNTVHIDLSALNPELVQAVQHVVIGPSSLIVHFNEVIGRGHFGCVYHGTLLDNDGKKIHCAVKSLNRITDIGEVSQFLTEGIIMKDFSHPNVLSLLGICLRSEGSPLVVLPYMKHGDLRNFIRNETHNPTVKDLIGFGLQVAKGMKYLASKKIVHRDLAARNCMLDEKFTVKVADFGLARDMYDKEYYSVHNKTGAKLPVKWMALESLQTQKFTTKSDVWSFGVLLWELMTRGAPPYPDVNTFDITVYLLQGRRLLQPEYCPDPLYEVMLKCWHPKAEMRPSFSELVSRISAIFSTFIGEHYVHVNATYVNVKCVAPYPSLLSSEDNADDEVDTRPASFWETS

Specifications

Accession Number NM_000245.2
Concentration 0.307 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 4233
Molecular Weight (g/mol) 78.753kDa
Name MET (F1200I) (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST affinity chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Immunogen KKRKQIKDLGSELVRYDARVHTPHLDRLVSARSVSPTTEMVSNESVDYRATFPEDQFPNSSQNGSCRQVQYPLTDMSPILTSGDSDISSPLLQNTVHIDLSALNPELVQAVQHVVIGPSSLIVHFNEVIGRGHFGCVYHGTLLDNDGKKIHCAVKSLNRITDIGEVSQFLTEGIIMKDFSHPNVLSLLGICLRSEGSPLVVLPYMKHGDLRNFIRNETHNPTVKDLIGFGLQVAKGMKYLASKKIVHRDLAARNCMLDEKFTVKVADFGLARDMYDKEYYSVHNKTGAKLPVKWMALESLQTQKFTTKSDVWSFGVLLWELMTRGAPPYPDVNTFDITVYLLQGRRLLQPEYCPDPLYEVMLKCWHPKAEMRPSFSELVSRISAIFSTFIGEHYVHVNATYVNVKCVAPYPSLLSSEDNADDEVDTRPASFWETS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias AUTS9/HGFR/RCCP2/c-Met
Common Name MET
Gene Symbol MET
Biological Activity 38pmol/μg x min
Species Baculovirus
Protein Tag None
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.