missing translation for 'onlineSavingsMsg'
Learn More

Human MESP1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18742997
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18742997 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18742997 Supplier R&D Systems Supplier No. MAB9219100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Mesp1 Monoclonal specifically detects Mesp1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesp1
Applications Western Blot, Immunocytochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 2030B
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BRJ9
Gene Alias BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1
Gene Symbols MESP1
Host Species Rabbit
Immunogen E. coli-dervived recombinant human MESP1, Met1-Gln85, Accession # Q9BRJ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 55897
Test Specificity Detects human MESP1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.