Learn More
Gibco™ Human Mesothelin Recombinant Protein, PeproTech®
Description
PeproTech's CHO cell-derived Recombinant Human Mesothelin is a glycoprotein containing 327 amino acid residues, and has a calculated molecular weight of approximately 36.4 kDa. As a result of glycosylation, Recombinant Human Mesothelin migrates with an apparent molecular mass of approximately 40-45 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | Q13421 |
| For Use With (Application) | Control, ELISA Standard, Binding Assay, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 10232 |
| Molecular Weight (g/mol) | 36.4 kDa |
| Name | Human Mesothelin |
| Purification Method | Purified |
| Quantity | 100 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.