missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mesothelin Propeptide/MPF Antibody, R&D Systems™
Goat Polyclonal Antibody
£418.32
Specifications
Antigen | Mesothelin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Mesothelin Polyclonal specifically detects Mesothelin in Human samples. It is validated for Western Blot.Specifications
Mesothelin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
MSLN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q13421 | |
10232 | |
Mouse myeloma cell line NS0-derived recombinant human Mesothelin Propeptide/MPF isoform 1 Arg35-Leu289 Accession # Q13421 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Mesothelin Propeptide/MPF in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant human Mesothelin is observed. |
For Research Use Only