missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mesenchymal Stem Cell Lineage Antibody Cocktail w/Isotype Control, BD
Brand: BD Biosciences 562530
Additional Details : Weight : 0.01200kg
Description
Host Species:
Clone: hMSC Negative Cocktail: 581, ICRF44, HIB19, HI30, G46-6
hMSC Isotype Control Negative Cocktail: X40, G155-178
Cross Reactivity Species: Hu
Flow Cytometry
Specifications
PE | |
Affinity Purified | |
50 Tests |