missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mer Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF891
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Mer Polyclonal specifically detects Mer in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Spezifikation
| Mer | |
| Polyclonal | |
| Biotin | |
| Q12866 | |
| MERTK | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Arg26-Ala499 Accession # Q12866 | |
| 50 μg | |
| Primary | |
| Detects human Mer in ELISAs and Western blots. In sandwich immunoassays, approximately 6% cross-reactivity with recombinant mouse Mer is observed, and less than 0.3% cross-reactivity with recombinant human (rh) Axl is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10461 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur