missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Meprin beta Subunit/MEP1B Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
£464.80
Specifications
Antigen | Meprin beta Subunit/MEP1B |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Description
Meprin beta Subunit/MEP1B Polyclonal specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Meprin beta Subunit/MEP1B | |
Western Blot 0.1 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta | |
MEP1B | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
NP_005916 | |
4225 | |
Mouse myeloma cell line NS0-derived recombinant human Meprin beta Subunit/MEP1B Leu21-Ser593 Accession # NP_005916 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Meprin beta Subunit/MEP1B in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse MEP1B is observed. |
For Research Use Only