missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Meprin alpha Subunit/MEP1A Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3220
This item is not returnable.
View return policy
Description
Meprin alpha Subunit/MEP1A Polyclonal specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Meprin alpha Subunit/MEP1A | |
| Polyclonal | |
| Unconjugated | |
| AAA21338 | |
| MEP1A | |
| Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A Val22-Gln601 Accession # AAA21338 | |
| 100 μg | |
| Primary | |
| Detects human Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse MEP1A and recombinant human MEP1B is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4224 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction